Learn More
Selleck Chemical LLC tetrathiomolybdate-E1166-25MG
SDP
Supplier:  Selleck Chemical LLC E116625MG
Encompass_Preferred
Tetrathiomolybdate (TM) is used in the clinic for the treatment of Wilson s disease by inducing dimerization of the metal-binding domain of the cellular copper efflux protein ATP7B (WLN4) through a unique sulfur-bridged Mo2S6O2 cluster
Catalog No. 50-313-7970