Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tetrodotoxin Monoclonal Antibody (3A10)
GREENER_CHOICE

Catalog No. PIMA549619 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549619 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549619 Supplier Invitrogen™ Supplier No. MA549619
  • $424.00
    $392.65 / Each of 1

    Save $31.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Tetrodotoxin is a potent neurotoxin that is found in a variety of organisms, including certain species of pufferfish, newts, and octopuses. It works by blocking voltage-gated sodium channels, which are responsible for the transmission of nerve impulses. This can lead to paralysis and death in animals that are exposed to it.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tetrodotoxin
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3A10
Concentration 6.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Maculotoxin; Spheroidine; Tarichatoxin; Tetrodotoxine
Host Species Mouse
Immunogen Conjugate of TTX hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.