Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetronic acid, 96%

Catalog No. p-7021660
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1407906 5 g
AAA1407914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1407906 Supplier Thermo Scientific Chemicals Supplier No. A1407906
Only null left
Add to Cart

CAS: 541-57-1 | C4H4O3 | 100.07 g/mol

Tetronic acid is used as a precursor of a variety of substituted and ring-fused furans and butenolides. It is an acidic building block for heterocyclic compounds for medicinal applications. It is used for selective O-alkyaltion reactions involving cesium fluoride. It is also used to prepare 3H-furo[3,4-b]quinolin-1-one by reacting with 2-amino-benzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetronic acid is used as a precursor of a variety of substituted and ring-fused furans and butenolides. It is an acidic building block for heterocyclic compounds for medicinal applications. It is used for selective O-alkyaltion reactions involving cesium fluoride. It is also used to prepare 3H-furo[3,4-b]quinolin-1-one by reacting with 2-amino-benzaldehyde.

Solubility
Soluble in water, dimethyl sulfoxide, acetonitrile and alcohol.

Notes
Incompatible with strong oxidizing agents and strong alkalis (bases).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-57-1
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00052187
InChI Key JZQBAGOECGRTSA-UHFFFAOYSA-N
SMILES OC1=CC(=O)OC1

Specifications

Melting Point ∽140°C (decomposition)
Assay Percent Range 96%
Beilstein 108442
Synonym tetronic acid, 2,4 3h,5h-furandione, furan-2,4 3h,5h-dione, tetrahydrofuran-2,4-dione, beta-tetronic acid, butanoic acid, 4-hydroxy-3-oxo-, .gamma.-lactone, tetronicacid, 2,4-furandione, tetrahydrofuran-3,5-dione, 3,5-dihydrofuran-2,4-dione
Solubility Information Soluble in water,dimethyl sulfoxide,acetonitrile and alcohol.
IUPAC Name 4-hydroxy-2,5-dihydrofuran-2-one
PubChem CID 521261
Formula Weight 100.07
Percent Purity 96%
Quantity 5 g
Chemical Name or Material Tetronic Acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-785-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.