Learn More
Apexbio Technology LLC TEV Protease 5000U
SDP
Supplier:  Apexbio Technology LLC K109810000
Encompass_Preferred
TEV protease is a highly specific cysteine protease with exceptional catalytic efficiency It recognizes the sequence ENLYFQ S but the amino acid at the P1 position can also be G A M C or H (1) TEV protease is commonly used to remove affinity purification tags from fusion proteins such as Glutathione S-transferase (GST) His or other tags As TEV protease itself carries a His-tag it can easily be removed by nickel affinity resin after the proteolysis reaction It is suitable for cleavage of His-tagged fusion proteins since incompletely cleaved His-tagged recombinant protein and the removed His-tagged portion along with His-tagged TEV protease itself can all be bound and removed through nickel column chromatography The desired target protein is obtained in the column flow-through in the meanwhile Overnight digestion at 4 with TEV protease is recommended TEV protease retains activity within pH 6 0-9 0 and is significantly reduced or even lost at pH 5 0 or below
Catalog No. 50-310-7263
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.