Learn More
Invitrogen™ TEX14 Polyclonal Antibody

Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (85%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TEX14 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TEX14 |
| Gene Accession No. | Q8IWB6 |
| Gene Alias | C85585; cancer/testis antigen 113; CT113; Inactive serine/threonine-protein kinase TEX14; LOW QUALITY PROTEIN: inactive serine/threonine-protein kinase TEX14; protein kinase-like protein SgK307; SGK307; Sugen kinase 307; testis expressed 14; testis expressed 14, intercellular bridge forming factor; testis expressed gene 14; testis expressed sequence 14; testis-expressed protein 14; Testis-expressed sequence 14; Testis-expressed sequence 14 protein; Tex14; Unknown (protein for MGC:133568) |
| Gene Symbols | TEX14 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.