Learn More
Aat Bioquest Texas Red-X succinimidyl ester Single isomer 178623-11-5 (5 mg)
SDP
Supplier:  Aat Bioquest 474
Encompass_Preferred
The amine-reactive Texas Red-X succinimidyl ester is a superior replacement to Texas Red Texas Red is the trademark of Invitrogen. This catalog is for 5 mg.
Catalog No. 50-224-9344
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.