Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Texwipe™ TexCaps Sterile Bottle Caps
SDP

Catalog No. 18366280 Shop All Texwipe Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
18-366-280 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-366-280 Supplier Texwipe™ Supplier No. TX001
This item is not returnable. View return policy

Designed to fit any 8, 16 or 32 oz. spray bottle with a 28/400 thread configuration. These caps offer a pouring option for disinfectants or cleaning solutions where there is a concern of contamination due to spray.

  • Flip-Top Cap has a no-touch design for the pouring spout eliminates potential contamination from glove contact
  • Use on any Texwipe™ 8, 16 or 32 oz. spray bottle product
  • Cap fits any 28/400 thread bottle
  • Contamination protection for a variety of operations
  • Purchase as needed, use to replace bottle spray triggers that may produce contamination
  • Gamma-irradiated according to AAMI guidelines to the sterility assurance level of 10-6
  • Tested for endotoxins
  • Double-bagged for introduction into sterile areas
  • Plastic case liner serves as a third bag, offering more protection for contamination prevention

Specifications

Product Type Flip-Top Bottle Cap
Sterility Sterile
Appearance Clear
Composition Polypropylene
Description 28/400 Thread
For Use With (Application) Pouring Disinfectant, Cleaning Solution or Water in Areas Where Spraying is a Concern
For Use With (Equipment) TX3265, TX3267, TX3270, TX3273, TX650, TX687, TX8270
Packaging Double Bagged
Quantity 48/Cs.
Volume (English) 8, 16, or 32 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.