Learn More
Valutek™ Textured Nylon Wiper, Cold Knife Cut
Catalog No. 191502022
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
12 x 12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-150-2022 12 x 12 in.
19-150-2027 9 x 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-150-2022 Supplier Valutek™ Supplier No. VTPNW1212
Add to Cart
Add to Cart
This item is not returnable. View return policy
Constructed from 100% continuous filament nylon in a double-knit, no-run, interlock pattern. This clean and absorbent wiper offers excellent softness and superior strength.
  • 100% continuous filament, double-knit nylon fiber
  • Cold knife cut edge
  • Excellent tensile strength, elasticity and durability
  • Low levels of particulate and extractable counts
  • Chemically compatible with common cleaning and disinfecting solutions
  • Laundered and packaged in a cleanroom, all Valutek™ wipers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 100-1000 (ISO 5-6) critical environment
  • The tough and abrasive surface of nylon makes it a great selection for cleaning stubborn contaminants
Specifications
Material Nylon
Dimensions (L x W) 12 x 12 in.
Sterility Non-sterile