Description
Apply directly to floor
Choose 30- or 60-layer mats.
- Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
- Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Specifications
| Color | Gray |
| Dimensions (L x W) | 46 x 36 in. |
| Quantity | 60 Sheets/Mat |