Learn More
Gerstel TF-SPME/TWISTER KIT PDMS/HLB
SDP
Supplier:  Gerstel KIT TFSPME HLB
Encompass
GERSTEL Thin Film Solid Phase Microextraction (TF-SPME)/Twister Starter Kit (PDMS/HLB)Consists of:2 x TF-SPME PDMS/HLB2 x GERSTEL PDMS Twisters2 x TF-SPME holders for 10/20 mL vials2 x 10 mL screw cap vials with caps and septa2 x 20 mL screw cap vials with caps and septa2 x TDU tube for Twister/TF-SPME1 x Instruction sheet
Catalog No. NC2584164
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.