Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFB2M Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540897
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540897 Supplier Invitrogen™ Supplier No. PA540897
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ECNPGPGILT QALLEAGAKV VALESDKTFI PHLESLGKNL DGKLRVIHCD Sequence homology: Horse: 100%; Human: 100%; Rabbit: 83%; Rat: 85%.

S-adenosyl-L-methionine-dependent methyltransferase which specifically dimethylates mitochondrial 12S rRNA at the conserved stem loop. Also required for basal transcription of mitochondrial DNA, probably via its interaction with POLRMT and TFAM. Stimulates transcription independently of the methyltransferase activity. Compared to TFB1M, it activates transcription of mitochondrial DNA more efficiently, while it has less methyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFB2M
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Tfb2m
Gene Accession No. Q9H5Q4
Gene Alias dimethyladenosine transferase 2, mitochondrial; FLJ22661; FLJ23182; HCV NS5A-transactivated protein 5; Hepatitis C virus NS5A-transactivated protein 5; Hkp1; h-mtTFB; h-mtTFB2; house-keeping protein 1; hTFB2M; mitochondrial 12S rRNA dimethylase 2; Mitochondrial transcription factor B2; mTFB2M; mtTFB2; NS5ATP5; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 2; TFB2M; transcription factor B2, mitochondrial
Gene Symbols Tfb2m
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TFB2M (aa 101-150).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64216
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.