Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFCP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 152031AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-0063


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

This gene encodes a transcription factor that binds the alpha-globin promoter and activates transcription of the alpha-globin gene. The encoded protein regulates erythroid gene expression, plays a role in the transcriptional switch of globin gene promoters, and it activates many other cellular and viral gene promoters. The gene product interacts with certain inflammatory response factors, and polymorphisms of this gene may be involved in the pathogenesis of Alzheimer′s disease.
Specifications

Specifications

TFCP2
Polyclonal
Unconjugated
TFCP2
CP2, LBP 1C, LSF, SAA3 enhancer factor, SEF, TFCP2, TFCP2C, transcription factor CP2, Transcription factor LSF
Rabbit
Antigen Affinity Chromatography
RUO
21422, 315309, 7024
-20°C
Liquid
Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q12800, Q9ERA0
Tfcp2
TFCP2 Fusion Protein Ag7353
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.