Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech TFF2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. p-7245684
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-8521 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-8521 Supplier PeproTech Supplier No. 500P312BT500UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: AEKPSPCQCS RLSPHNRTNC GFPGITSDQC FDNGCCFDSS VTGVPWCFHP LPKQESDQCV MEVSDRRNCG YPGISPEECA SRKCCFSNFI FEVPWCFFPK SVEDCHY. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TFF-2. Anti-Human TFF-2-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human TFF-2 by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti- Human TFF-2 (500-P312) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TFF-2. Western Blot: To detect Human TFF-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible development reagents the detection limit for Recombinant Human TFF-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P312BT-1MG will be provided as 2 x 500 μg

Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing and protection of the gastrointestinal epithelia. These peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain. SP (spasmolytic polypeptide), also designated Trefoil factor 2 (TFF2) precursor, is a trefoil protein that functions to inhibit gastrointestinal motility and gastric acid secretion. SP may also act as a structural component of the gastric mucus, possibly by stabilizing glycoproteins in the mucus gel through interactions with carbohydrate side chains. A downregulation of SP expression is associated with primary gastric cancer, and a progressive loss of this protein is likely to be involved in the early stage of the multi-step gastric carcinogenesis pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFF2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene Tff2
Gene Accession No. Q03403
Gene Alias mSP; pancreatic spasmolytic polypeptide; PSP; SML1; SP; spasmolysin; Spasmolytic polypeptide; spasmolytic protein 1; Tff2; Trefoil factor 2; trefoil factor 2 (spasmolytic protein 1)
Gene Symbols Tff2
Host Species Rabbit
Immunogen E.coli-derived, 12.0 kDa Recombinant Human TFF-2.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7032
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.