Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFG Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7141451
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2364 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2364 Supplier Bethyl Laboratories, Inc Supplier No. A302342AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

TFG protein, a recently identified novel protein, is a human homolog of mouse TFG protein. It contains an N-terminal coiled-coil structure, a SPYGQ-rich region followed by octicosapeptide repeats. Research shows that TFG can modulate SHP-1 activity and is a novel member of the NF-kB pathway. The physiological functions of this protein remain largely elusive, but TFG is thought to function by associating with other proteins, including TANK and NEMO. TFG was first identified as a partner of NTRK1 and is involved in generating the thyroid TRK-T3 oncogene, as well as oncogenic rearrangements with ALK in anaplastic lymphoma and NOR1 in mixoid chondrosarcoma.

Specifications

Antigen TFG
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TFG
Gene Accession No. Q92734
Gene Alias TFG
Gene Symbols TFG
Host Species Rabbit
Immunogen Between 175 and 225
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10342, 21787
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.