Learn More
AdipoGen TFMU
SDP
Supplier:  AdipoGen CDXT0003GG25
Encompass_Preferred
Chemical. CAS 575-03-1. Formula C10H5F3O3. MW 230.14. Synthetic. This compound is widely used as a substrate for Cytochrome P450. It has also been used as a marker substrate for assessment of Drug-Drug interactions to multiple isoforms of UDP-glucuronosyltransferases. Suitable as pH-indicator. Trifluoromethylumbelliferone is a slightly longer wavelength analog of 4-methylcoumarin 4-MU with a pKa that more closely matches physiological pH values.
Catalog No. 50-256-0053
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.