Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TfR (Transferrin R) Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10092243
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10092243 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10092243 Supplier R&D Systems Supplier No. NB10092243

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody has been used in 12 publications

TfR (Transferrin R) Polyclonal specifically detects TfR (Transferrin R) in Human, Mouse, Rat samples. It is validated for Block/Neutralize, IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay, Western Blot.

Specifications

Antigen TfR (Transferrin R)
Applications Blocking Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay, Western Blot
Classification Polyclonal
Concentration 0.558 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:50-1:200, Proximity Ligation Assay reported in scientific literature (PMID 31091436)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P02786
Gene Alias CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Gene Symbols TFRC
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Transferrin R (between amino acids 20-75) [UniProt P02786]
Molecular Weight of Antigen 89 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7037
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.