Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat TfR (Transferrin R) Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FB10079U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB10079U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB10079U100 Supplier R&D Systems Supplier No. FAB10079U100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen TfR (Transferrin R)
Applications Flow Cytometry
Classification Monoclonal
Clone OX26
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Host Species Mouse
Immunogen PHA activated rat lymphocytes
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7037
Test Specificity Detects rat TfR in Flow Cytometry.
Target Species Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.