Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TFR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542733 Supplier Invitrogen™ Supplier No. PA542733
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YVSLDNAVLG DDKFHAKTSP LLTSLIESVL KQVDSPNHSG QTLYEQVVFT Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.

This gene is a member of the transferrin receptor-like family and encodes a single-pass type II membrane protein with a protease associated (PA) domain, an M28 peptidase domain and a transferrin receptor-like dimerization domain. This protein mediates cellular uptake of transferrin-bound iron and mutations in this gene have been associated with hereditary hemochromatosis type III. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFR2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Tfr2
Gene Accession No. Q9UP52
Gene Alias HFE3; MGC126368; TfR2; TFRC2; transferrin receptor 2; transferrin receptor protein 2; Trfr2
Gene Symbols Tfr2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TFR2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7036
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.