Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TG 003
GREENER_CHOICE

Catalog No. 433610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-361-0 10 mg
43-365-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-361-0 Supplier Tocris Bioscience™ Supplier No. 4336/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent inhibitor of Clk-family kinases; also inhibits DYRK1A/B

Potent, ATP-competitive inhibitor of Clk-family kinases (IC50 values are 15, 20 and 200 nM for mClk4, 1 and 2 respectively and >10 μM for mClk3). Also inhibits DYRK1A/B (IC50 values are 24 and 34 nM respectively). Suppresses serine/arginine-rich protein phosphorylation; affects the regulation of alternative splicing by phosphorylation of SR protein both in vitro and in vivo.

Chemical Identifiers

CAS 719277-26-6
Molecular Formula C13H15NO2S

Specifications

Quantity 10 mg
Formula Weight 249.33
Purity >98%
Molecular Weight (g/mol) 249.328
Chemical Name or Material TG 003
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.