Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGE Virus Monoclonal Antibody (1Q17)

Catalog No. 01670336
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-336 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-336 Supplier Invitrogen™ Supplier No. MA17602
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7602 detects TGE from virus samples. MA1-7602 has been successfully used in ELISA, and Western blot procedures. The MA1-7602 immunogen is a purified TGE virus 1E11.

TGE (Porcine transmissible gastroenteritis coronavirus (strain Purdue) (TGEV)) is a replicase polyprotein of coronaviruses. Replicase polyprotein is a multifunctional protein: it contains the activities necessary for the transcription of negative stranded RNA, leader RNA, subgenomic mRNAs and progeny virion RNA as well as proteinases responsible for the cleavage of the polyprotein into functional products.[UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGE Virus
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1Q17
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias Transmissible Gastro Enteritis
Host Species Mouse
Immunogen Purified TGE virus 1E11.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.