Learn More
Sino Biological Mouse Latent TGF-beta 1 / TGFB1 Protein (His Tag), Biotinylated 100ug
Supplier:  Sino Biological 50698M08HB100
Encompass_Preferred
A DNA sequence encoding the mouse Latent TGFB1 (P04202) (Met1-Ser390) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6259
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.