Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGFB2 Rabbit anti-Human, Mouse, Polyclonal, liquid, OriGene
SDP

Catalog No. 501684759
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4759 Supplier OriGene Supplier No. TA890046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transforming growth factor type beta 2 (TGFB2) is a member of the TGF-beta family of cytokines, which are ubiquitous, multifunctional, and essential for survival. They play important roles in growth and development, inflammation and repair, and host immunity for many different cell types. Human TGFB2 consists of two disulfide-linked, identical subunits each of which is a single-chain polypeptide of 112 residues.

Specifications

Antigen TGFB2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TGFB2
Gene Accession No. P27090, P61812
Gene Alias G-TSF, LDS4, TGF-beta2
Gene Symbols TGFB2
Host Species Rabbit
Immunogen Recombit protein of human TGFB2
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21808, 7042
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.