Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TGFBR1 Recombinant Rabbit Monoclonal Antibody (PSH14-14)
GREENER_CHOICE

Catalog No. MA562931
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA562931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA562931 Supplier Invitrogen™ Supplier No. MA562931
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Positive Control: THP-1 cell lysate, MCF7 cell lysate, SH-SY5Y cell lysate, Neuro-2a cell lysate, RAW264.7 cell lysate, C6 cell lysate. Tissue Specificity: Low tissue specificity Subcellular Location: Cell membrane, Cell junction, tight junction, Cell surface, Membrane raft. Sequence Similarities: 97.5% Mouse/Rat Predicted band size: 56 kDa.

TGFBR1 is a transmembrane serine/threonine kinase in the TGFBR1 superfamily, which also includes ACVRs and BMPRs. Upon binding TGF-b, TGFBR2 dimerizes with and activates TGFBR1 via phosphorylation leading to downstream phosphorylation of SMADs by TGFBR1. TGFBR1 forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF-beta signal from the cell surface to the cytoplasm. The encoded protein is a serine/threonine protein kinase. Mutations in this gene have been associated with Loeys-Dietz aortic aneurysm syndrome (LDAS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGFBR1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone PSH14-14
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol, 0.1% BSA and 0.2% ProClin 950; pH 7.4
Gene TGFBR1
Gene Accession No. P36897, P80204, Q64729
Gene Alias AAT5; activin A receptor type II-like kinase, 53kDa; activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ACVRLK4; ALK5; Alk-5; AU017191; ESK2; ESS1; LDS1; LDS1A; LDS2A; MSSE; mutant transforming growth factor beta receptor I; ser; serine/threonine-protein kinase receptor R4; SKR4; TbetaRI; tbetaR-I; tgf beta receptor 1; TGF-beta receptor type I; TGF-beta receptor type-1; TGF-beta type I receptor; TGFBR1; TGFR1; TGFR-1; Transfor; transforming growth factor beta receptor 1; transforming growth factor beta receptor I; transforming growth factor, beta receptor 1; transforming growth factor, beta receptor I; transforming growth factor, beta receptor I (activin A receptor type II-like kinase, 53kDa); transforming growth factor-beta receptor type I
Gene Symbols TGFBR1
Host Species Rabbit
Immunogen Recombinant protein within Human TGF beta Receptor I aa 34-503.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21812, 29591, 7046
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.