Learn More
Invitrogen™ TGIF Monoclonal Antibody (OTI1B12)

Description
TGIF Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | TGIF |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1B12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TGIF1 |
| Gene Accession No. | Q15583 |
| Gene Alias | 5'-TG-3'-interacting factor 1; AA959811; AI462167; Homeobox protein TGIF1; HPE4; TALE family homeobox; TALE homeobox TG-interacting factor; Tfig; TG interacting factor 1; TGFB induced factor homeobox 1; TGFB-induced factor homeobox 1; TGIF; Tgif1; transforming growth factor-beta-induced factor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.