Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TGN46 Mouse anti-Human, Porcine, Rat, Clone: 1F6D5, Proteintech
SDP

Catalog No. 501736900 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6900 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6900 Supplier Proteintech Group Inc Supplier No. 664771IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.

Specifications

Antigen TGN46
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1F6D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TGOLN2
Gene Accession No. O43493
Gene Alias TGN38, TGN38 homolog, TGN46, TGN48, TGN51, TGOLN2, trans Golgi network protein 2, TTGN2
Gene Symbols TGOLN2
Host Species Mouse
Immunogen TGN46 Fusion Protein ag4470
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100519476, 10618, 192152
Target Species Human, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.