Learn More
Selleck Chemical LLC TH1760-E1274-25MG
SDP
Supplier:  Selleck Chemical LLC E127425MG
Encompass_Preferred
TH1760 (Compound 18) is a first-in-class potent selective and cell-active NUDT15 (MTH2) inhibitor with an IC50 value of 25 nM
Catalog No. 50-313-8027