Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thalidomide 4'-oxyacetamide-alkylC4-amine
Description
Thalidomide 4'-oxyacetamide-alkylC4-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a short alkyl linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Thalidomide - linker 3
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | PROTAC Toolbox |
| CAS | 2245697-86-1 |
| Synonym | Thalidomide - linker 3 |
| Quantity | 25 mg |
| Molecular Formula | C19H22N4O6.HCl |
| Formula Weight | 438.86 |
| Purity | 95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?