Learn More
Eurofins Discoverx Thapsigargin, 1 mg. (92-1148)
Supplier:  Eurofins Discoverx 921148
Encompass
Thapsigargin is a naturally occurring, cell-permeable plant sesquiterpene lactone that non-competitively inhibits the sarco/endoplasmic reticulum calcium ATPase (SERCA, IC50 ~2-80 nM). Effectively increases intracellular calcium levels upon SERCA inhibition, leading to cell activation, increased histamine release from mast cells, and increased cancer cell proliferation. Acts as an anti-inflammatory and induces autophagy in-vivo. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9679
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.