Learn More
Cayman Chemical Thapsigargin, 67526-95-8, 1 mg
SDP
Supplier:  Cayman Chemical 105221
Encompass_Preferred
A non-competitive, cell permeable inhibitor of calcium transport by SERCAs (IC50 values are cell type-dependent and range from ~2-80 nM); increases intracellular calcium, leading to cell activation, histamine release, and increased cell proliferation; has anti-inflammatory and anti-cancer effects in vivo
Catalog No. NC1412406
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.