Learn More
AdipoGen Thapsigargin (high purity),Chemical. CAS: 67526-95-8
SDP
Supplier:  AdipoGen AGCN20003M005
Encompass_Preferred
Thapsigargin (high purity), AdipoGen Life Sciences. Chemical. CAS: 67526-95-8. Formula: C34H50O12. MW: 650.8. Isolated from Thapsia garganica. Intracellular Ca2+ signaling probe. Specific and sensitive inhibitor of SERCA. Non-TPA/PMA tumor promoter. Histamine release inducer. Apoptosis inducer. Mitochondrial dysfunction inducer. NOS modulator. Angiogenesis inhibitor. Stimulator of arachidonic acid metabolism in macrophages. Autophagy inducer. TRAIL sensitizer.
Catalog No. 50-464-294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.