Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thaumatin, Thaumatococcus daniellii

Catalog No. AAJ6366803
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6366803 1 g
AAJ63668MC 100 mg
AAJ63668MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6366803 Supplier Thermo Scientific Chemicals Supplier No. J6366803
Add to Cart

A mixture of the intensely sweet proteins thaumatin I and thaumatin II | CAS: 53850-34-3

Thaumatin is used as a mixture of the intensely sweet proteins, used to enhance the taste of various foods, Flavoring Agents, Food Additives, Sweetening Agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thaumatin is used as a mixture of the intensely sweet proteins, used to enhance the taste of various foods, Flavoring Agents, Food Additives, Sweetening Agents.

Solubility
Soluble in water (25 mg/ml), and 60% ethanol. Insoluble in ether, and benzene .

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53850-34-3
MDL Number MFCD00132420

Specifications

Color Brown to Yellow
Quantity 1 g
Merck Index 14,9273
Solubility Information Soluble in water (25mg/ml),and 60% ethanol. Insoluble in ether,and benzene .
Physical Form Powder
Chemical Name or Material Thaumatin, Thaumatococcus daniellii
EINECSNumber 258-822-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.