Learn More
AdipoGen Thaxtomin A
SDP
Supplier:  AdipoGen BVT0206M005
Encompass_Preferred
Chemical. CAS 122380-18-1. Formula C22H22N4O6. MW 438.4. Isolated from Streptomyces bottropensis Gö-Dra 17. Diketopiperazine antibiotic. Phytotoxin. Herbicide. Natural cellulose synthesis inhibitor. Plant cell necrosis inducer. Induces common scab disease of potato.
Catalog No. 50-256-2061
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.