Learn More
Description
Specifications
Specifications
| Description | similar to thrombospondin 1 precursor |
| Cross Reactivity | Monkey |
| Target Species | Monkey |
| For Use With (Application) | Quantitative |
| Gene ID (Entrez) | 705413 |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Storage Requirements | Store at 4°C for 6 months or at -20°C for 12 months. Avoid repeated freeze-thaw cycles. Centrifuge tubes briefly to spin down all components to the bottom before opening. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.