Learn More
Bachem Lactoferricin B25 Trifluoroacetate
SDP
Supplier:  Bachem 4087382.0001
Encompass_Preferred
Lactoferricin B25 Trifluoroacetate, H-Phe-Lys-Cys-Arg-Arg-Trp-Gln-Trp-Arg-Met-Lys-Lys-Leu-Gly-Ala-Pro-Ser-Ile-Thr-Cys-Val-Arg-Arg-Ala-Phe-OH, 146897-68-9, C141H224N46O29S3, Mr 3123.82, 1mg. LFcinB 25, Lactoferricin B (17-41) (bovine). The antimicrobial peptide lactoferricin B25 is a fragment of cow's milk protein lactoferrin. The peptide exhibits potent dose-dependent and time-dependent anticancer activity against human stomach cancer cell cultures via apoptosis and autophagy. LFcinB25 is expected to yield novel anticancer drugs.
Catalog No. 50-194-6675
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.