Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beach Lane Books The Beak Book by Robin Page
SCI_ED

Catalog No. S000953 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Format:
Paperback Book
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S000953 Paperback Book
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000953 Supplier Beach Lane Books Supplier No. 9781534460416
Add to Cart
Add to Cart

Literature for educational purposes.

  • Educational book for learning use

Specifications

Grade Pre-K+
Format Paperback Book
Product Type Paperback, Birds, Observation
Subject Birds
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.