Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSI, LLC: The Case of the Kidnapped Cookies Kit
SCI_ED

Catalog No. p-4365890 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Product Type:
the Case of the Kidnapped Cookies Lab Activity Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S06630 the Case of the Kidnapped Cookies Lab Activity Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S06630 Supplier Aqua Phoenix Scientific Supplier No. CRI5303KIT
This item is not returnable. View return policy

Includes

teacher's guide, student worksheets and casebooks and a rubric

Who stole the cookies from the cookie jar?

  • Activity involves investigating the theft of Otis Spunkmeyer cookies
  • Students need to figure out which of their staff members took the warm, chewy cookies
  • Clues involve a ransom note written in Spanish, cut from several different local newspapers, with red stain (possibly lipstick?) and white powder and soil left near the note
  • Students to determine whose DNA is on the envelope the note was found in
  • Activity also involves trial for culprit
  • Guilty party pays for free cookies for everyone as sentence
  • For middle school students
  • Class size: 30 students
  • Requires a microscope (sold separately)

Specifications

For Use With (Application) For middle school students
Includes Teacher's guide, student worksheets and casebooks
Requires Microscope
Class Size 30 Students
Product Type the Case of the Kidnapped Cookies Lab Activity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.