Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSI, LLC: The Case of the Missing Mascot Kit
SCI_ED

Catalog No. p-4365895 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Product Type:
the Case of the Missing Mascot Lab Activity Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S06628 the Case of the Missing Mascot Lab Activity Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S06628 Supplier Aqua Phoenix Scientific Supplier No. CRI5301KIT
This item is not returnable. View return policy

Includes

A teacher's guide, student worksheets and casebooks

Who took the mascot?

  • Activity involves investigating the theft of the school mascot
  • Clues include a note, some strange-looking leaves, a mouth guard and some white fibers left at the scene
  • Students will have to figure out whether it was taken by one of the rival schools before the big game
  • Investigations include analyzing DNA results, using dichotomous keys to identify selected plants, analyzing handwriting, fingerprints, and bite marks, and a fiber flame test
  • A trial is staged after finding the culprit
  • For middle school students
  • Class size: 30 students

Specifications

Product Type the Case of the Missing Mascot Lab Activity Kit
For Use With (Application) For middle school students
Includes Teacher's guide, student worksheets and casebooks
Class Size 30 Students
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.