Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ The Chemistry Of Invisible Inks Stem Kit
SDPSCI_ED

Catalog No. S00662 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S00662 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00662 Supplier United Scientific™ Supplier No. AISINKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The The Chemistry Of Invisible Inks Stem Kit allows students to take what they learn in the activities and design their own activities.

  • STEM based curriculum kits accent the scientific method and independent inquiry, featuring hands-on learning materials and exciting interactive digital content.
  • Kits contain structured, guided, and open investigations, at introductory, intermediate, and advanced levels, to make inquiry-based learning affordable and fun.
  • This kit is a unique teaching tool for introducing your students to the fascinating chemistry and physics behind invisible inks, i.e. colorless substances usable for writing that can be made to run color using a chemical or physical process.
  • Students investigate how organic fluids and chemical reagents can be used as invisible inks.
  • Relying on historical example, students create various invisible inks and security documents for other student groups to analyze and decode!
  • Includes materials for two detailed activities (each with a 'model' and 'inquiry' investigation paths - a total of four investigational activities).
  • Includes enough materials for up to 40 students, as well as digital content including teacher and student guides.
  • Covered concepts include: data analysis, chemical and physical processes, elements, mixtures and compounds, pH indicators, transmitted vs. reflected light, the periodic table, fluorescent compounds, types of chemical reactions, and hypothesis testing.

Specifications

Grade 6 to 12
Includes DVD with PDF Teacher and Student Guides
Class Size 40 Students
Product Type STEM Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.