Learn More
Bachem (Gln22)-Amyloid Beta Protein (1-40) Trifluoroacetate
SDP
Supplier:  Bachem 4039928.0500
Encompass_Preferred
(Gln22)-Amyloid Beta Protein (1-40) Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Gln-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH, 144410-00-4, C194H296N54O57S, Mr 4328.88, 0.5mg. Amyloid b-Protein (1-40) E22Q, Amyloid b-Protein (1-40) Dutch Mutation, [Gln22] b-Amyloid (1-40). The Dutch mutation (E22Q) of amyloid b-peptide aggregates more readily than the wild-type peptide and the resulting fibrils show increased neurotoxicity. The mutant peptide E22Q induced apoptosis of cerebral endothelial cells at a concentration of 25 μm, whereas WT Ab 1-40 and the Italian mutant E22K showed no effect.
Catalog No. 50-194-6607
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.