Learn More
Bachem (Gly22)-Amyloid Beta Protein (1-40) Trifluoroacetate
SDP
Supplier:  Bachem 4035372.1000
Encompass_Preferred
(Gly22)-Amyloid Beta Protein (1-40) Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Gly-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH, 175010-18-1, C191H291N53O56S, Mr 4257.80, 1mg. Amyloid b-Protein (1-40) E22G, Amyloid b-Protein (1-40) Arctic Mutation, [Gly22] b-Amyloid (1-40). The highly neurotoxic arctic mutant (E22G) of Ab has been used to study the mechanisms underlying the formation of soluble and insoluble b-amyloid aggregates. As the wild-type Ab, the arctic mutant preferably assembles in the presence of GM1 ganglioside.
Catalog No. 50-194-6608
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.