Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harper Collins The Last Straw: Kids Vs. Plastics by Susan Hood and Christiane Engel
SCI_ED

Catalog No. S000945 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Subject:
Pollution, Recycling
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Subject
S000945 Pollution, Recycling
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000945 Supplier Harper Collins Supplier No. 9780062981394
Add to Cart
Add to Cart

Literature for educational purposes

  • Educational Book

Specifications

Grade Pre-K+
Format Paperback Book
Product Type Paperback, Environmental
Subject Pollution, Recycling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.