Learn More
Johannes Lieder The Mouth Parts of Insects
SCI_ED
Catalog No. S02809 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Catalog No. Mounting Type
S02809 Cross and Longitudinal Section
1 options
Catalog No. S02809 Supplier Johannes Lieder Supplier No. 5570
5570 The Mouth Parts of Insects, 20 microscope slides
  • 5570 The Mouth Parts of Insects
  • 20 Microscope Slides
  • Periplaneta, cockroach, chewing biting mouth parts, dissected w.m.
  • Carabus, ground beetle, biting mouth parts of a carnivore showing extraintestinal digestion, w.m.
  • Gomphocerus, grasshopper, biting mouth parts of a herbivore w.m.
  • Vespa vulgaris, wasp, biting mouth parts of a carnivore w.m.
  • Melolontha, cockchafer, chewing mouth parts of a herbivore, dissected and w.m.
  • Apis mellifica, honey bee, leaking sucking mouth parts of worker w.m.
  • Apis mellifica, honey bee, t.s. of mouth parts
  • Pieris, butterfly, sucking mouth parts w.m.
  • Pieris, butterfly, t.s. of mouth parts
  • Pyrrhocoris, bug, piercing sucking mouth parts w.m.
  • Pyrrhocoris, bug, t.s. of mouth parts
  • Culex pipiens, common mosquito, piercing sucking mouth parts of female w.m.
  • Culex pipiens, common mosquito, mouth parts of male w.m.
  • Culex pipiens, t.s. of female mouth parts
  • Stomoxys or Tabanus, stable fly, piercing sucking mouth parts w.m.
  • Tabanus, t.s. of mouth parts
  • Musca domestica, house fly, leaking sucking mouth parts w.m.
  • Musca domestica, house fly, t.s. of mouth parts
  • Bombyx mori, silkworm moth, chewing mouth parts w.m.
  • Curculionidae, weevil, head with mouth parts w.m.
Specifications
Classification Zoology
Material Elastin Stained Glass
Mounting Type Cross and Longitudinal Section
Type Microscope Slides
Slide Type Prepared Slide