Learn More
Perkin Elmer US LLC Near Infrared Reflectance Accessory (NIRA) for Frontier, External Mounted
Supplier:  Perkin Elmer US LLC L128401N
Encompass_Preferred
The Near Infrared Reflectance Accessory (NIRA) is designed for diffuse reflectance measurements of light-scattering solids, liquids, and powders using the Frontier FT-NIR or FT-IR/NIR spectrometer.
Catalog No. 50-201-6502
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.