Learn More
Bachem (Des-Glu22)-Amyloid Beta Protein (1-40) Trifluoroacetate
SDP
Supplier:  Bachem 4091431.1000
Encompass_Preferred
(Des-Glu22)-Amyloid Beta Protein (1-40) Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH, 1678416-36-8, C189H288N52O55S, Mr 4200.75, 1mg. Amyloid b-Protein (1-40) Osaka or Japanese Mutation, Amyloid b-Protein (1-40) E22D. The Osaka mutation was the first deletion-type mutation to be identified in APP and Ab. The Ab E22delta mutant is more resistant to degradation by two major Ab-degrading enzymes, neprilysin and insulin-degrading enzyme. Synthetic mutant Ab showed unusual aggregation properties with enhanced oligomerization but no fibrillization. It also inhibited hippocampal long-term potentiation more efficiently than wild-type Ab. A transgenic mouse model containing APP with the E693delta mutation has been developed. APP(OSK)-Tg mice exhibit intraneuronal Ab E22delta oligomers and memory impairment as early as eight months of age.
Catalog No. 50-194-6605
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.