Learn More
Bachem (Ala1)-PAR-4 (1-6) amide (mouse) Trifluoroacetate
SDP
Supplier:  Bachem 4035529.0001
Encompass_Preferred
(Ala1)-PAR-4 (1-6) amide (mouse) Trifluoroacetate, H-Ala-Tyr-Pro-Gly-Lys-Phe-NH2, 352017-71-1, C34H48N8O7, Mr 680.80, 1mg. (Ala1)-Thrombin Receptor-Like 3 (1-6) amide (mouse), (Ala1)-Proteinase Activated Receptor 4 (1-6) amide (mouse), (Ala1)-Coagulation Factor II Receptor-Like 3 (1-6) amide (mouse), AYPGKFamide, AYPGKF-NH2 Selective Protease-Activated Receptor 4 (PAR4) Agonist. The peptide AYPGKF-amide based on the proteolytically-revealed tethered ligand sequence of the murine proteinase-activated receptor-4 (PAR-4) was shown to cause platelet aggregation with an EC50 value of about 15 uM.
Catalog No. 50-194-6788
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.