Learn More
Perkin Elmer US LLC Rhodamine 101 Fluorescence Standard
Supplier:  Perkin Elmer US LLC N4202027
Encompass_Preferred
The Rhodamine 100 Standard is a mixture of Polyol (>90%) and Rhodamine B (<10%). It is packaged in a cuvette with a diagonal cutoff and flame-sealed. ISO-certified standard.
Catalog No. 50-201-6328
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.