Learn More
Bachem Retrocyclin-1 Trifluoroacetate
SDP
Supplier:  Bachem 4045698.1000
Encompass_Preferred
Retrocyclin-1 Trifluoroacetate, Cyclo(-Gly-Ile-Cys-Arg-Cys-Ile-Cys-Gly-Arg-Gly-Ile-Cys-Arg-Cys-Ile-Cys-Gly-Arg), 724760-19-4, C74H128N30O18S6, Mr 1918.42, 1mg. RC-100. The theta-defensin retrocyclin-1, also called RC-100, is a circular octadecapeptide containing an internal trisulfide ladder. Besides its antimicrobial activity, retrocyclin-1 has a remarkable ability to protect human cells in vitro from infection by T- and M-tropic strains of HIV-1. Its effectiveness against HIV-1 results from the prevention of viral entry, and is correlated with its ability to recognize and bind with high affinity to glycoproteins and glycolipids implicated in HIV-1 entry, such as gp120 (KD = 35.4 nM), CD4 (KD = 31 nM), and galactosylceramide (KD = 24.1 nM). Retrocyclin represents a new class of small molecule HIV-1 entry inhibitors.
Catalog No. 50-194-6658
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.