Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

The Young Scientists Club™ Kit 5: Phases of Matter
SCI_ED

Catalog No. S02364
Change view
Click to view available options
Kit 5: Phases of Matter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S02364 Kit 5: Phases of Matter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD (1,2). Small parts. Not for children under 3 yrs.
Catalog No. S02364 Supplier The Young Scientists Club™ Supplier No. SS9251105
CHOKING HAZARD (1,2). Small parts. Not for children under 3 yrs.

Includes

10 bags of raisins, 10 balloons, 10 Ziploc™ bags, 10 Popsicle™ sticks, 10 funnels, 10 bags of Borax™ powder, 10 bottles of glue, 10 fizzy tablets, 10 manuals and 10 student manuals/teacher guides

Students study the three phases of matter.

  • Students mix solids and liquids to create gas, a bouncy ball and goop
  • These fun experiments will excite any budding scientist
  • Kit size: 30 students

Winner! Dr. Toy Best 100 products, The National Parenting Center Seal of Approval, Learning Research Institute Outstanding Resource Award, National Parenting Publications Honors Award, Learning Magazine Teacher's Choice Award, Practical Homeschooling Reader Award.

Specifications

Product Type Phases of Matter Classroom Kit
For Use With (Application) For students study the three phases of matter by mixing solids and liquids to create gas, a bouncy ball, and goop
Includes 10 bags of raisins, 10 balloons, 10 ziploc™ bags, 10 popsicle™ sticks, 10 funnels, 10 bags of borax™ powder, 10 bottles of glue, 10 fizzy tablets, 10 manuals, and 10 student manuals/teacher guides
Class Size 30 Students
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.