Learn More
AdipoGen Theacrine, 2309-49-1, MFCD00042781, 1 g
SDP
Supplier:  AdipoGen AGCN20466G001
Encompass_Preferred
MF: C9H12N4O3, MW: 224.2, ≥98%, Appearance: White to off-white solid. Purine alkaloid related to caffeine. Potent antioxidant. Not through direct scavenging of ROS but strengthen antioxidant systems in vivo. Anti-inflammatory and antinociceptive compound. Suggested to have adenosine receptor agonistic activity and also acting on dopamine receptors. Shown to have potent sedative and hypnotic properties. Ameliorated learning and memory impairments caused by central fatigue. Shown to inhibit cAMP-specific PDE4 and cGMP-specific PDE5 activity in vivo and in vitro. Increased glucose level, decreased lactic acid concentration, reduced LDH activity and elevated the expressions of both GLUT1/3 in restraint-stressed mice. Can increase the solubility of aromatic systems in water.
Catalog No. 50-114-6365
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.