Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Theanine, Amino Acid Analog, Solid, 5 g

Catalog No. 502706819
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6819 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6819 Supplier Abcam Supplier No. AB1411875G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Neuroprotective and cognitive-enhancing amino acid analog of glutamine.

  • Molecular Weight: 174.2 Da
  • Chemical Description: Amino acid analog of glutamine, commonly found in green tea
  • Biological Activity: Binds to AMPA, kainate, NMDA and group I mGlu receptors; displays neuroprotective and cognitive enhancing effects; modifies self-renewal properties of cancer stem cells
  • Properties: Blood-brain barrier permeable and active in vivo
  • Form: Solid
  • CAS Number: 3081-61-6
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule AMPA, Kainate, NMDA, Group I mGlu Receptors
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Formulation 100% (2S)-2-amino-4-(ethylcarbamoyl)butanoic acid
Quantity 5 g
For Use With (Application) Neuroprotection, cognitive enhancement, cancer stem cell research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.